Learn More
Medchemexpress LLC HY-P0079 10mg Medchemexpress, Neuromedin N CAS:92169-45-4 Purity:>98%
Supplier: Medchemexpress LLC HYP007910MG
Medchemexpress, HY-P0079 10mg Neuromedin N CAS:92169-45-4 Neuromedin N is a potent modulator of dopamine D2 receptor agonist binding in rat neostriatal membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.