Learn More
Walter Products™ Neuron Model
Shop All Walter Products Products
Click to view available options
:
Neuron Model; Dimensions (L x W x H): 17 x 13 x 5 in.; Material: PVC
Description
- Neuron model displays the cell body, and the myelin sheath with nodes of Ranvier
- Schwann cells can be revealed by removing a part of the dendrite
- Mounted on base
- Corresponding key card is included
Specifications
Specifications
Product Type | Anatomical Model |
Dimensions (L x W x H) | 17 x 13 x 5 in. |
Material | PVC |