Learn More
Enzo Life Sciences Neutralizing reagent, (5ml)
SDP
Supplier:  Enzo Life Sciences ADI801475
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Neutralizing buffer for low pH solutions; for use with cyclic nucleotide and isoprostane kits. Formulation: Liquid. A high pH buffer to neutralize 0.1M hydrogen chloride in an EIA system; for use with cyclic nucleotide and isoprostane kits. Long Term Storage: Ambient.
Catalog No. 50-200-6817
missing translation for 'QuantityLeft'