Learn More
Multimedia Lessons for Interactive Whiteboards: Earth Science



The perfect complement to any earth science curriculum — use on the whiteboard for the entire class or on a computer for an individual student.
Supplier: NewPath Learning 546308
Description
Key Features:
- Virtual Lab Investigation: Allows students to experiment, perform simulations and draw conclusions
- Custom Tool Bar: Allows students to draw, write and add arrows and text blocks to customize the lessons by adding details to any screen
- Lesson Preparation: Consists of a series of narrated, visual presentations, each featuring highly detailed graphics and illuminating animations
- Interactive Activities: Includes a variety of interactive activities, a virtual lab investigation and a Lab Activity Guide; allows students to learn concepts, apply knowledge to other modules and develop skills to perform the virtual lab investigation
- Visual Resources: Customize lessons using additional images, arrows, text and callouts provided in the Custom Tool Bar
- Vocabulary: Illustrated vocabulary; show or hide definitions or play audio version
- Assessment: Use in Quiz Mode, Practice Mode or Play Mode
Lesson Topics
Identifying Minerals - What is a Mineral?, Properties of Minerals, Identifying Minerals, How Do Minerals Form?, Mineral Resources, Uses of Minerals; Virtual Lab Investigation: Identifying Minerals
Rocks - Classifying Rocks, Igneous Rocks, Sedimentary Rocks, Metamorphic Rocks, Coral Reefs, The Rock Cycle; Virtual Lab Investigation: Identifying Rocks
Plate Tectonics - Earth's Interior, Types of Heat Transfer, Continental Drift, Sea-Floor Spreading, Theory of Plate Tectonics; Virtual Lab Investigation: Plate Tectonics Simulation
Earthquakes - Types of Stress, Kinds of Faults, How Earth's Surface Changes, Types of Seismic Waves, Measuring Earthquakes, Earthquake Damage and Safety; Virtual Lab Investigation: Modeling Earthquakes
Volcanoes - Volcanoes and Plate Boundaries, Properties of Magma, Volcanic Eruptions, Life Cycle of a Volcano, Volcanic Landforms, Hot Springs and Geysers; Virtual Lab Investigation: Investigating Volcanoes
Earth's Surface - Landforms, Topographic Maps, Weathering & Erosion, Mechanical Weathering, Chemical Weathering, Cycle of Erosion and Deposition, Soil; Virtual Lab Investigation: Effect of Chemical Weathering on Rocks
Earth's Atmosphere and Weather - The Atmosphere, Weather Factors, Clouds, Weather Patterns, Predicting the Weather; Virtual Lab Investigation: Forecasting the Weather
Earth's Climate - What is Climate?, Causes of Climate, Climate Regions/Types of Climate, Causes of Climate Change, Short-Term Climate Change, Global Warming, Ozone Depletion; Virtual Lab Investigation: Investigating Climate Change
Our Solar System - Formation of the Solar System, Geocentric and Heliocentric Systems, The Sun, The Inner Planets, The Outer Planets, Comets, Asteroids and Meteors; Virtual Lab Investigation: Exploring Our Solar System
Sun-Earth-Moon System - How Earth Moves; Seasons on Earth; Gravity and Motion; Phases, Eclipses and Tides; Earth's Moon; Missions to the Moon; Virtual Lab Investigation: Simulating the Sun-Earth-Moon System
Order Info
Lessons available with single-user or site license. Single-user license can be installed and used on a single computer/Interactive Whiteboard. Site license permits access to any computer/Interactive Whiteboard within a single school building.
Certifications
Accredited by SMART™ and Promethean™ for use on their Interactive Whiteboards.
Specifications
8 | |
Earth's Climate Multimedia Lesson | |
Windows™, Mac™ | |
CD-ROM | |
Smart™, Promethean™ | |
5 Modules, digital inquiry based student activity guide |
6 | |
Interactive whiteboard or computer | |
Earth's Climate Multimedia Lesson | |
Single User | |
Multisensory learning, perfect resource for interactive whiteboards and the lessons are also ideal for individual student instruction on any computer | |
Earth Science |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.