Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NewPath Learning™ Safety Flip Chart



Mounted on a sturdy easel.
Supplier: NewPath Learning™ 348001
Description
- 10 double-sided, laminated 12 × 18 in. (30.5 × 45.7cm) charts
- Side 1 features colorful, graphic overview of product
- Side 2 serves as “write-on/wipe-off” activity chart featuring questions, labeling exercises, vocabulary review and more
- Activity Guide features black-line copy-masters and exercises
Lab safety:
Safety Symbols and Labels; Personal Protective Equipment; Safety in the Lab; Hygiene Practices in the Lab; Chemical Exposure; Chemical Handling; Eye Safety; Chemical Spills; Bloodborne Pathogens Safety; Biological Safety
Specifications
Curriculum Mastery Flip Chart Set | |
Laminated | |
Photosynthesis, modern genetics, osmosis and diffusion, microlife bacteria, fungi and protists, rocks, minerals, plate tectonics, elements and the periodic table, chemical reactions, properties of atoms |
8 | |
Science | |
Academic Standard for California, Florida, National, New York, Texas |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction