Learn More
NEXTAL BIOTECHNOLOGIES NeXtal DWBlock Cations Suite
Supplier:  NEXTAL BIOTECHNOLOGIES 130908
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
NeXtal Cations Suite for protein crystallization. 96 unique conditions in 1.5 ML Deep Well Block format.
Catalog No. 50-197-8938
missing translation for 'QuantityLeft'