Learn More
NEXTAL BIOTECHNOLOGIES NeXtal DWBlock Cryos Suite
Supplier:  NEXTAL BIOTECHNOLOGIES 130903
Encompass_Preferred
NeXtal Cryos Suite for protein crystallization. 96 unique conditions in 1.5 ML Deep Well Block format.
Catalog No. 50-197-8933
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.