Learn More
NEXTAL BIOTECHNOLOGIES NeXtal Tubes MBClass II Suite
Supplier:  NEXTAL BIOTECHNOLOGIES 130712
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
NeXtal MBClass II Suite for protein crystallization. 96 unique conditions in 10 ml tube format.
Catalog No. 50-197-8919