Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF 023, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
A subtype-selective, competitive and reversible P2X1 receptor antagonist. Displays IC50 values of 0.21, 28.9, > 50 and > 100 μM for human P2X1, P2X3, P2X2, and P2X4-mediated responses respectively. Selective over adrenoceptors, histamine and P2Y receptors. Also selectively inhibits the α-subunit of Go/i (EC50 ∽300 nM). Also inhibits DNA-binding activity of HMGA2 (IC50 = 10.63 μM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 1162.86 |
Percent Purity | >98% |
Chemical Name or Material | NF 023 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction