Learn More
Gold Biotechnology Inc Nickel Agarose Beads (High Density), 50 mL
Supplier: Gold Biotechnology Inc H32050
Store at 4°C. Nickel Agarose Resin is available in bulk or in prepacked columns. It has a binding capacity of >25 mg/ml with approximately 60 kDa protein. In addition, it allows for purification of proteins under native or denaturing conditions. GoldBio Nickel Agarose works very well with the His-Tag Buffer Set.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.