Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nickel (II) Sulfate, Hexahydrate
SCI_ED

Catalog No. p-4416416 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Color Blue-Green
Quantity 500 g
Assay Percent Range >95%
Linear Formula NiSO4·6H2O
Formula Weight 262.87
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Nickel (II) Sulfate, Hexahydrate
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.