Learn More
Gold Biotechnology Inc Nickel NTA Agarose Beads, 10 mL
SDP
Supplier:  Gold Biotechnology Inc H35010
Encompass_Preferred
H-350-10 Nickel-NTA Agarose Resin has a binding capacity of ~50 mg/ml. In addition, it allows for purification of proteins under native or denaturing conditions. GoldBio Nickel Agarose works very well with the His-Tag Buffer Set.
Catalog No. 50-153-2797
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.