Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™

Catalog No. p-7083076 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5g
25 g

Serves as a coenzyme of the dehydrogenases. β-Nicotinamide Adenine Dinucleotide, MP Biomedicals is one of the biologically active forms of nicotinic acid that occurs in living cells primarily in the oxidized state. It is a major electron acceptor mol

  • Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
  • Acts as a coenzyme for hydride-transfer enzymes
  • Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
  • Recently reported to be a P2Y11 agonist

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H26N7O14P2-
Molecular Weight (g/mol) 662.422
InChI Key BAWFJGJZGIEFAR-NNYOXOHSSA-M
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=O)N

Specifications

pH 2 to 4 (1% aq. soln.)
Quantity 5g
Solubility Information Soluble in water (200mg/mL)
Formula Weight 663.40 (anhydrous)
Physical Form White Powder
Percent Purity ≥93%
Recommended Storage −20°C, desiccate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.