Learn More
Electron Microscopy Sciences NIGHTSEA™ Fluorescence Adapter VI Light Head, Royal Blue
Supplier: Electron Microscopy Sciences SFALHRB

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing individual light head, barrier filter, and viewing shield combinations. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. The barrier filter clicks on to the ring adapter magnetically, so it is easy to remove it to switch back to white light viewing.
Color: Royal Blue
Excitation: 440-460 nm
Emission filter: 500 nm longpass
Fluorophores: GFP, eGFP, fluorescein
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.