Learn More
Medchemexpress LLC HY-14284 10mg Medchemexpress, Nilvadipine CAS:75530-68-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1428410MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-14284 10mg Nilvadipine CAS:75530-68-6 Nilvadipine is a potent calcium channel antagonist, and the IC50 value is around 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1582
missing translation for 'QuantityLeft'