Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitenpyram, Thermo Scientific™

Catalog No. p-7214477
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 150824-47-8
Molecular Formula C11H15ClN4O2
Molecular Weight (g/mol) 270.72
InChI Key CFRPSFYHXJZSBI-DHZHZOJOSA-N
IUPAC Name [(1E)-1-{[(6-chloropyridin-3-yl)methyl](ethyl)amino}-2-nitroethenyl](methyl)amine
SMILES CCN(CC1=CC=C(Cl)N=C1)C(\NC)=C\[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 270.72
Melting Point 70.0°C to 84.0°C
Flash Point >70°C
Solubility Soluble in water; Insoluble in methanol and dimethyl sulfoxide
Chemical Name or Material Nitenpyram
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.