Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrate Nitrogen Standard, 1000 ppm N (4427 ppm NO3-), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
Quantity | 120 mL |
CAS Min % | 97.0, 0.7 |
CAS Max % | 100.0, 0.73 |
Color | Colorless |
Concentration or Composition Notes | 1000ppm N (4427ppm NO3-) |
Equivalence | 1 mL = 1 mg N |
Physical Form | Liquid |
Chemical Name or Material | Nitrate IC Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction