Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 50% (v/v), Ricca Chemical
SDP

Catalog No. p-3252000 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Safety-coated Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7697-37-2, 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Concentration 50% (v/v)
CAS Min % 49.0
CAS Max % 51.0
Packaging Safety-coated Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Nitric Acid, 50% (v/v)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.