Learn More
StatLab™ Nitrical Decalcifier
SDP
A safe and gentle standard AFIP formulation.
Supplier:  StatLab™ 12131
 View more versions of this product
Catalog No. STL12131
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
This item is not returnable. View return policy
Description
Description
  • Made from semiconductor grade acid and highly purified reagent grade water
  • Very mild solution that works rapidly with great results
  • Can be used for all types of specimens from bone marrow to cortical bone
Specifications
Specifications
4 x 1 gal.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.