Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell BioClean-D™ Sterile Disposable Hood

Catalog No. 19668342 Shop All Ansell Products
Click to view available options
:
Sterile hood
This item is not returnable. View return policy
This item is not returnable. View return policy

The BioClean-D™ hood is a sterile single use hood designed and constructed using three sections for a perfect fit. Made from antistatic durable low-linting CleanTough™ material, the hood features an elasticated face-opening with reinforced edges for exceptional comfort and protection

  • ISO Class 4 and EU GMP Grade A compatible
  • Sterilized by Gamma irradiation
  • Elasticated face opening with reinforced edges
  • Low linting and anti-static property fabric
  • Universal size
  • Low particulate
  • ASTM D6978 05 TESTED
  • HELMKE DRUM TESTED
Applications: Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, industrial

Compliance

ANSI/ISEA Z87.1

Specifications

Size Universal
Color White
Disposable Yes
For Use With (Application) Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, industrial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.