Learn More
Ansell™ BioClean™ Durableu™ High Durability Cleanroom Overshoes
Shop All Ansell Products
Click to view available options
:
Size: 18 in.; Blue
Description
- High Tensile Strength
- Slip-Resistant
- Resists tears and abrasions
- Non-Woven spunbonded Polypropylene/cast polyethylene laminate
- Manufactured in a facility holding ISO 9001:2008
- In accordance with ISO 2859
Specifications
Specifications
Size | Universal |
Material | Polyethylene, Polypropylene |
Color | Blue |
Certifications/Compliance | ISO 9001:2015 |
Packaging Quantity | 50 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; eight outer bags per lined carton (400 pieces) |