Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, 100.0%, 99.0% purity, For GC analysis, MP Biomedicals™

Catalog No. p-4405155 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mL
1 L

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym benzene, nitro, nitro-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Color Yellow
Density 1.196g/mL at 25°C
Assay Percent Range 99%
Quantity 500 mL
Formula Weight 123.12
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene, For GC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.