Learn More
Perkin Elmer US LLC NM FID Tower 250 Dry Hydrogen Generator, 250 mL/min
Supplier:  Perkin Elmer US LLC N9300011
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
With a maximum output capacity of 250 ml/min, one generator can supply up to 6 GC's. The compact design allows the generator to be installed directly in the laboratory eliminating the requirement for long gas lines and guaranteeing the delivery of high purity gas to your GC. A sophisticated control system connected to a touch screen control, continuously monitors the vital operating parameters to ensure a safe and consistent performance. Built in sensors will shut the generator down if internal/external leaks are present, contaminated water, low water or over pressure. The generators meet the strict safety guidelines to be certified for CE, MET & FCC. Maximum Pressure: 11 bar/160 psi Purity: 99.9996%
Catalog No. 50-209-3365
missing translation for 'QuantityLeft'