Learn More
Electron Microscopy Sciences Numbered Gunshot Residue Sampler
Supplier: Electron Microscopy Sciences 7648016
Numbered, certified aluminum mount with a double-coated adhesive carbon tab applied to secure the sample in a clean, glass vial 25 mm OD x 55 mm high (1" x 2.17"). Packaged in a box of 100. Numbers can be specified as 100 consecutive numbers between 0 and 9999 (greatest numbers). Please inform customer service about the numbering sequence when ordering this product.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.