Learn More
Ace Glass, Inc. Column, 4" ID x 18" , 4" O-ring grooved flat flange top, #50 Ace-Thred bottom
Supplier:  Ace Glass, Inc. 586245
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
4 I.D. x 18 chromatography column with a size 4 o-ring grooved flat flange top and #50 Ace-Thred bottom. Supplied with silicone o-ring.
Catalog No. 50-972-395
missing translation for 'QuantityLeft'