Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nomifensine, Tocris Bioscience™

Catalog No. 199250 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent noradrenalin and dopamine uptake inhibitor. Antidepressant

Nomifensine is a potent inhibitor of noradrenaline and dopamine uptake (Ki values are 4.7, 26 and 4000 nM for inhibition of noradrenaline, dopamine and 5-HT uptake respectively, in rat brain). Increases central dopamine levels in rats following systemic administration. Displays antidepressant activity.

Chemical Identifiers

CAS 32795-47-4
Molecular Formula C20H22N2O4
Molecular Weight (g/mol) 354.406
InChI Key GEOCVSMCLVIOEV-VIEYUMQNSA-N
Synonym unii-76s8cuh5mr component
PubChem CID 13396304
IUPAC Name (Z)-but-2-enedioic acid;(4R)-2-methyl-4-phenyl-3,4-dihydro-1H-isoquinolin-8-amine
SMILES CN1CC(C2=C(C1)C(=CC=C2)N)C3=CC=CC=C3.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 354.4
Percent Purity >98%
Chemical Name or Material Nomifensine
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.