Learn More
US Biomax Inc Normal colon tissue array with colon cancer tissue as control, including TNM, clinical stage and pathology grade, 24 cases/ 72 cores, unstained slide
SDP
Supplier:  US Biomax Inc CO727
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Normal colon tissue microarray, containing 19 cases of normal colon tissue, 2 cases of cancer adjacent normal colon tissue, plus 3 cases of colon adenocarcinoma, triplicate cores per case
Catalog No. NC3359114
missing translation for 'QuantityLeft'