Learn More
US Biomax Inc SMN601 : Normal small intestine tissue array, including TNM, clinical stage and pathology grade, 30 cases/ 60 cores
Normal small intestine tissue microarray, containing 7 cases of normal duodenum tissue, 12 cases of normal small intestine tissue, plus 3 cases of adenocarcinoma, duplicate cores per case