Learn More
Honeywell™ North™ First Aid Kit Refills
Catalog No. p-118343
Click to view available options
Product Type:
This item is not returnable. View return policy
This item is not returnable. View return policy
Use these refill items to restock Fisher Scientific™ first aid kits and unitized North first aid kits
Rugged, reverse-tuck, color-coded packs permit quick identification. Clear plastic packaging lets color code show through, protects the pack from moisture, and includes a tear flap for quick opening.
  • Yellow: bandages
  • Blue: antiseptics
  • Green: special items such as insect sting or poison plant treatment, or miscellaneous items
  • Orange: personal protection (for products such as gloves)
Compliance
Each pack is stamped with lot number and expiration date (if applicable), in compliance with FDA requirements, and shows graphic instructions to assist in proper use of the item.
Specifications
For Use With (Application)
Unitized first aid kits
Product Type
PVP Iodine Wipes
Certifications/Compliance
FDA