Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nortriptyline Hydrochloride, MP Biomedicals™

Catalog No. p-4679697 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 894-71-3
Molecular Formula C19H22ClN
Molecular Weight (g/mol) 299.84
MDL Number MFCD00058024
InChI Key SHAYBENGXDALFF-UHFFFAOYSA-N
Synonym nortriptyline hydrochloride, pamelor, allegron, altilev, nortrilen, desmethylamitriptyline hydrochloride, acetexa, psychostyl, sensival, vividyl
PubChem CID 441358
IUPAC Name hydrogen methyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CNCCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Color White
Quantity 250 mg
Formula Weight 299.8
Physical Form Solid
Chemical Name or Material Nortriptyline Hydrochloride
EINECSNumber 212-973-0
storageNote1 Protect from light.
Recommended Storage Store at room temperature (15° to 30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.