Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nortriptyline Hydrochloride, USP, 97-101.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157904 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

N1054, 894-71-3, C19H21N•HCl

Spectrum™ Chemical Nortriptyline Hydrochloride, USP is a tricyclic antidepressant used to treat nocturnal bedwetting. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 894-71-3
Molecular Formula C19H22ClN
Molecular Weight (g/mol) 299.84
MDL Number MFCD00058024
InChI Key SHAYBENGXDALFF-UHFFFAOYSA-N
IUPAC Name hydrogen methyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CNCCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 97 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.