Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ NOVAX™ Class 0 Natural Rubber Electrical Insulating Gloves - 11 in.
Shop All Protective Industrial Products Inc. Products
Click to view available options
Color:
Black
Orange
Size:
8
9
10
Description
- Class 0 electrical insulating gloves constructed of natural rubber
- Flexible material allows workers to easily handle small, delicate parts
- Contoured shape is comfortable and helps to reduce hand fatigue
- Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
- Ideal for use around electrical and arc flash hazards
- Individually numbered for traceability
- Stamped with factory manufacturing date
- Silicone-free
Specifications
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 0, Type I, NFPA 70E |
Applications | Industrial |
Color | Black |
Exterior Finish | No Texture |
Material | Natural Rubber |
Size | 8 |
For Use With (Application) | Electrical Utility Work, Hybrid Automobiles, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Series | 150-0-11 |
Length (English) | 11 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction