Learn More
Invitrogen™ Novex™ 10% Zymogram Plus (Gelatin) Protein Gels, 1.0 mm

Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol.
$226.65
Specifications
Gel Size | Mini |
---|---|
Product Line | Novex™ |
Gel Percentage | 0.1 |
Gel Type | Zymogram Gel |
Mode of Separation | Molecular Weight |
Catalog Number | Mfr. No. | No. of Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Wells | Price | Quantity & Availability | |||||
ZY00100BOX
![]() ![]() |
Invitrogen™
ZY00100BOX |
10-well |
Pack of 10
for $226.65
Save 3% Reg : |
|
|||||
ZY00105BOX
![]() ![]() |
Invitrogen™
ZY00105BOX |
15-well |
Pack of 10
for $226.65
Save 3% Reg : |
|
|||||
Description
Novex 10% Zymogram Plus (Gelatin) gels are useful for the detection and characterization of proteases that use gelatin as a substrate. The proteases are run under denaturing conditions and visualized as clear bands against a dark background using a simple renaturing, developing, and staining protocol. Zymogram gels are commonly used to detect matrix metalloproteinases. Novex 10% Zymogram Plus gels are highly sensitive, detecting as little as 5 x 10-6 units of collagenase.
Formulation
Novex Zymogram Plus gels are Tris-glycine gels with 0.1% gelatin incorporated into the gel. They are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water, and casein or gelatin as the substrate. All have a 4% stacking gel.
Choose the right gel format for your experiments
Novex 10% Zymogram Plus gels come in a fixed concentration of 10% gelatin and three different well formats, including 10-, 12- and 15-well.
Recommended buffers
Use Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC2675) with these gels. Novex Zymogram Renaturing Buffer (LC2670) and Novex Zymogram Developing Buffer (LC2671) are also recommended.
Specifications
Mini | |
0.1 | |
Molecular Weight | |
30 to 150 kDa (Tris-Glycine Buffer) | |
Store at 2°C to 8°C. Do not freeze. | |
8 cm |
Novex™ | |
Zymogram Gel | |
10 Gels/Box | |
Wet Ice | |
1.0 mm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.