Learn More
TARGETMOL CHEMICALS INC NPS-2143 HYDROCHLORIDE 10MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NPS-2143 hydrochloride (NPS-2143 HCL) is a selective potent calcium ion-sensing receptor antagonist. purity: 99%