Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NS 19504
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
NS 19504 is a selective activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1) (EC50 = 11 μM). Activates BK channels in native smooth muscle cells from guinea pig bladder and reduces spontaneous phasic contractions in bladder strips ex vivo. Displays no effect on contractions induced by a high K+ concentration or at 65 other receptors tested, including L- and N- type Ca2+ channels, Nav, KATP and hERG channels.
Specifications
Specifications
Chemical Name or Material | 5-[(4-Bromophenyl)methyl]-2-thiazolamine |
CAS | 327062-46-4 |
Quantity | 10 mg |
Target | Ca2+-Activated Potassium Channel Activators |
Molecular Formula | C10H9BrN2S |
Formula Weight | 269.16 |
Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction