Learn More
Medchemexpress LLC HY-108586 10mg Medchemexpress, NS3623 CAS:343630-41-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10858610MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-108586 10mg NS3623 CAS:343630-41-1 NS3623 is an activator of human ether-a-go-go-related gene (hERG1/KV11.1) potassium channels. NS3623 activates the IKr and Ito currents and has antiarrhythmic effect. NS3623 has a dual mode of action, being an inhibitor of hERG1 channels[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8582
missing translation for 'QuantityLeft'