Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NU 7026, Tocris Bioscience™

Catalog No. p-4784221 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective DNA-PK inhibitor

ATP-competitive inhibitor of DNA-dependent protein kinase (DNA-PK). Displays selectivity over other PIKK family enzymes (IC50 values are 0.23, 13.0, > 100 and > 100 μM for DNA-PK, PI3K, ATM and ATR respectively). Radiosensitizes both proliferating and quiescent mouse embryonic fibroblast cells to IR and inhibits DSB repair.

Chemical Identifiers

CAS 154447-35-5
Molecular Formula C17H15NO3
Molecular Weight (g/mol) 281.311
InChI Key KKTZALUTXUZPSN-UHFFFAOYSA-N
Synonym dna-pk inhibitor ii, 2-morpholin-4-yl-benzo h chromen-4-one, 2-morpholino-4h-benzo h chromen-4-one, 2-4-morpholinyl-4h-naphtho 1,2-b pyran-4-one, 2-4-morpholinyl-4h-naphthol 1,2-b pyran-4-one, 2-morpholin-4-ylbenzo h chromen-4-one, dna-dependent protein kinase inhibitor ii, 2-morpholin-4-yl benzo h chromen-4-one, d03tvf, d0w1st
PubChem CID 9860529
IUPAC Name 2-morpholin-4-ylbenzo[h]chromen-4-one
SMILES C1COCCN1C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3

Specifications

Quantity 10 mg
Formula Weight 281.31
Percent Purity >98%
Chemical Name or Material NU 7026
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.