Learn More
ACROBiosystems resDetect™ GENIUS™ Nuclease ELISA Kit (Residue Testing)
Supplier: ACROBiosystems CRSA03196TESTS
resDetect™ GENIUS™ Nuclease ELISA Kit is based on ELISA sandwich method and designed for quantitative determination the residue of nuclease in biologicals. It contains Nuclease and a pair of antibodies against the eneyme, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of nuclease concentrations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.