Learn More
NUP188 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody
$154.46
Specifications
Antigen | NUP188 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2311
![]() |
Bethyl Laboratories, Inc
A302322AT |
20 μL |
Each of 1 for $154.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
May function as a component of the nuclear pore complex (NPC).Specifications
NUP188 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Nucleoporin NUP188 homolog, hNup188, NUP188, KIAA0169 | |
NUP188 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5SRE5 | |
23511 | |
Between 1600 and 1650 | |
Primary | |
4° C | |
NUP188 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.