Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Nutrient Broth
Shop All AquaPhoenix Scientific Products

Click to view available options
Quantity:
100 g
500 g
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | Beige |
Form | Powder |
Manufacturing Origin | United States |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction