Learn More
Bachem o-Conotoxin GVIA Trifluoroacetate
SDP
Supplier:  Bachem 4016306.0500
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
o-Conotoxin GVIA Trifluoroacetate, H-Cys-Lys-Ser-Hyp-Gly-Ser-Ser-Cys-Ser-Hyp-Thr-Ser-Tyr-Asn-Cys-Cys-Arg-Ser-Cys-Asn-Hyp-Tyr-Thr-Lys-Arg-Cys-Tyr-NH2, 106375-28-4, C120H182N38O43S6, Mr 3037.39, 0.5mg. o-Conotoxin GVIA from the venom of the fish-hunting cone snail Conus geographus efficiently blocks neuronal calcium channels. This presynaptic inhibition of calcium influx prevents the voltage-activated release of acetylcholine.
Catalog No. 50-260-105
missing translation for 'QuantityLeft'