Learn More
Medchemexpress LLC HY-101094 5mg Medchemexpress, Ocaperidone CAS:129029-23-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010945MG
Encompass_Preferred
Medchemexpress, HY-101094 5mg Ocaperidone CAS:129029-23-8 Ocaperidone is an effective antipsychotic agent, acting as a potent 5-HT 2 and dopamine D 2 antagonist, and a 5-HT 1A agonist, with K i s of 0.14 nM, 0.46 nM, 0.75 nM, 1.6 nM and 5.4 nM for 5-HT 2 , a 1 -adrenergic receptor, dopamine D 2 , histamine H 1 and a 2 -adrenergic receptor, respectively, and a p EC 50 and pK i of 7.60 and 8.08 for h5-HT 1A . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2035
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.