Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Octanoylcarnitine chloride, Tocris Bioscience™

Catalog No. 060550 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Intermediate in lipid metabolism

Homolog of acetylcarnitine chloride (Cat. No. 0355). Acylcarnitines are important intermediates in lipid metabolism.

Chemical Identifiers

CAS 18822-86-1
Molecular Formula C15H30ClNO4
Molecular Weight (g/mol) 323.858
InChI Key MYMFUYYXIJGKKU-UHFFFAOYSA-N
Synonym octanoyl-carnitine, octanoyl carnitine hydrochloride, +/--octanoylcarnitine chloride, octanoylcarnitine chloride, rac octanoyl carnitine chloride, octanoyl-dl-carnitine hydrochloride, 3-carboxy-2-octanoyloxy propyl trimethylazanium chloride, octanoyl-dl-carnitin chloride, a+/--octanoylcarnitine chloride, octanoic acid, ester with 3-carboxy-2-hydroxypropyl trimethylammonium chloride
PubChem CID 177508
IUPAC Name (3-carboxy-2-octanoyloxypropyl)-trimethylazanium;chloride
SMILES CCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Specifications

Quantity 50 mg
Formula Weight 323.86
Percent Purity >98%
Chemical Name or Material (±)-Octanoylcarnitine chloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.