Learn More
Showa Denko America Inc OHpak LB-806M
Supplier:  Showa Denko America Inc F6429202
Encompass_Preferred
SEC, aqueous GPC polyhydroxymethacrylate packed columns for aqueous SEC (GFC) analysis, low column bleeding allows its use with light scattering detectors, the eluent can be replaced with DMF enabling the analysis of polar polymers, fulfills USP L38 and L39 requirements, dimensions: 8.0 mm ID x 300 mm L, target MW range: 500 − 20,000,000 (estimated maximum) Daltons in pullulan
Catalog No. 50-161-3278
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.