Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl Acetate 60.0+%, TCI America™
SDP

Catalog No. p-7117139 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 693-80-1
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
MDL Number MFCD00056326
InChI Key GYGAZRPDUOHMAF-KHPPLWFESA-N
Synonym Acetic Acid Oleyl Ester
PubChem CID 5363289
IUPAC Name (9Z)-octadec-9-en-1-yl acetate
SMILES CCCCCCCC\C=C/CCCCCCCCOC(C)=O

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 mL
Formula Weight 310.52
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleyl Acetate
EINECSNumber (2)-0762
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.