Learn More
Cytiva Nanosep Centrifugal Devices with Omega™ Membrane - 3K, gray (24/pkg)
Supplier:  Cytiva OD003C33
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Simple, Reliable Concentrating and Desalting of 50 to 500 μL Samples-Rapidly concentrates 50-500 μL sample volumes to 15 μL-Achieve 50X concentration and > 90% recovery in just minutes-Available with low proteinbinding Omega™ and Bio-Inert-A wide range of MWCOs, color-coded for easy identification-Constructed of low-binding polypropylene -Ultrasonically welded seals prevent bypass or seal failure-Fits standard centrifuge rotors that accept 1.5 mL tubes This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9590