Learn More
Dunlop™ Onguard™ Steel Midsole Work Boots
SDP
Ideal for demolition crews and recycling plants
Supplier:  Dunlop™ 868021300
 View more versions of this product
Catalog No. 19-024516
This item is not returnable. View return policy
Description
Description
  • Polyblend PVC construction provides moderate chemical protection
  • Full steel midsole for complete underfoot protection
  • Steel toe for optimal impact and crush resistance
  • Feature Ultragrip™ sole, pull-up and kick-off lugs, and replaceable insoles
  • 16 in. high (41cm)
  • Waterproof
  • Black
Compliance
Stainless-steel midsole meets or exceed ASTM™ F2413-11 for puncture resistance and impact and crush resistance. Steel midsole and toe also meet CSA Z195 Grade 1.
Specifications
Specifications
ASTM
Demolition crews and recycling plants
Polyblend
Ultragrip™ sole, full Steel midsole
Steel
Black
16 in.
13
Standard
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.