Learn More
Glass Expansion OpalMist DC Nebulizer 1.0mL/min, high purity PFA, compatible with Thermo Q, RQ and TQ.
Supplier: Glass Expansion A311PFA1
OpalMist DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.5mm ID x 1.6mm OD x 700mm long tubing (1 pc) & DC Gas Fitting Type 31 with ratchet connector (1 pc). Compatible with Thermo Q, RQ and TQ.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-16-50 & 501907289.Recommended nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.