Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Open Shelf Storage
SCI_ED

Allows for a clear view for all items stored. The Open Shelf Storage has a height of 84 inches and comes in a variety of widths and depths.

Supplier:  Diversified Spaces™ OS1413K

 View more versions of this product

Catalog No. S47434


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • 84 in. Height
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Dowel construction that is reinforced with glue and screws
  • Open shelving with one fixed and four adjustable shelves
  • Adjustable shelf weight capacity is 40 lbs. per square foot
  • The unit is finished with a chemical resistant, earth-friendly UV finish
  • Made in the USA

Warranty and Services

Lifetime Limited Warranty

Specifications

Specifications

Open Shelf Cabinet
4 (Adjustable); 1 (Fixed)
Natural
12 in.
84 in.
48 in.
Black rubber base molding
Red Oak
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.