Learn More
Beckman Coulter OptiMATE 80% Iodixanol, 500 mL
Supplier:  Beckman Coulter D01358
Encompass
Iodixanol for your purification needs. Ideal for step gradients common to viral, organelle, EV, and protein separations. Concentrated to 80%, OptiMATE Iodixanol can enable you to increase your sample per tube and decrease your bottle-related waste.Volume: 500mL, Density: 1.420 ± 0.004 g/mL, Concentration: 80% w/v, Refractive Index: 1.460 ± 0.004, Endotoxin Level: ≤1 EU/mL, Sterile: Yes (autoclaved), Shelf Life: 18 months, Bottle Dimensions: 17.6 cm x 7.2 cm / 6.93" x 2.83" (H x W)
Catalog No. 50-274-3744
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.